Learn More
Thermo Scientific™ PepMap™ 300 C4 HPLC Columns
Use this nanocolumn, packed with 300Å wide pore silica for high-resolution analyses of hydrophobic proteins and peptides.
$902.78 - $1,788.35
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
DX163579
|
Thermo Scientific™
163579 |
5 μm | 0.075 mm | 150 mm |
Each for $1,479.97
|
|
DX163591
|
Thermo Scientific™
163591 |
5 μm | 0.3 mm | 5 mm |
Pack of 5 Each for $902.78
|
|
DX164720
|
Thermo Scientific™
164720 |
5 μm | 0.3 mm | 150 mm |
Each for $1,752.83
|
|
DX164721
|
Thermo Scientific™
164721 |
5 μm | 1 mm | 150 mm |
Each for $1,788.35
|
|
Description
Separate hydrophobic peptides and proteins, providing higher recoveries with Thermo Scientific™ Acclaim™ PepMap™ 300 C4 LC Columns.
- High resolution in protein identification, biomarker discovery and systems biology
- Particular loadability allows hghest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
Specifications
3% | |
Yes | |
300 Å |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.