Please login to your online account to display your discounted pricing

PEPA, Tocris

Allosteric potentiator of AMPA receptor desensitization

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material PEPA
Assay Percent Range >99%
Molecular Formula C16H16F2N2O4S2
Formula Weight 402.44
Synonym 2-[2, 6-Difluoro-4-[[2-[(phenylsulfonyl)amino]ethyl]thio]phenoxy]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

292910

 
r & d systems
RDS292910
10mg Each for $188.60

292950

 
r & d systems
RDS292950
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 402.44
Quantity 10mg
Chemical Name or Material PEPA
CAS 141286-78-4
Molecular Formula C16H16F2N2O4S2
Assay Percent Range >99%
Synonym 2-[2, 6-Difluoro-4-[[2-[(phenylsulfonyl)amino]ethyl]thio]phenoxy]acetamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature