missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentylmagnesium chloride, 1.3M solution in THF, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436218000

 View more versions of this product

Catalog No. AC436218000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pentylmagnesium chloride
−17°C
0.967
Undesignated
6393-56-2,109-99-9
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf
JFWWQYKSQVMLQU-UHFFFAOYSA-M
magnesium;pentane;chloride
10953547
Liquid
0.9670g/mL
AcroSeal™ Glass Bottle
65.0°C
800mL
C5H11ClMg
Solubility in water: reacts.
CCCC[CH2-].[Mg+2].[Cl-]
130.9
130.9

Chemical Identifiers

6393-56-2
130.9
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf
magnesium;pentane;chloride
C5H11ClMg
JFWWQYKSQVMLQU-UHFFFAOYSA-M
10953547
CCCC[CH2-].[Mg+2].[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

SDS Documents