missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentylmagnesium chloride, 1.3M solution in THF, AcroSeal™, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436211000
Specifications
Pentylmagnesium chloride | |
−17°C | |
0.967 | |
Undesignated | |
6393-56-2,109-99-9 | |
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf | |
JFWWQYKSQVMLQU-UHFFFAOYSA-M | |
magnesium;pentane;chloride | |
10953547 | |
Liquid |
0.9670g/mL | |
AcroSeal™ Glass Bottle | |
65.0°C | |
100mL | |
C5H11ClMg | |
Solubility in water: reacts. | |
CCCC[CH2-].[Mg+2].[Cl-] | |
130.9 | |
130.9 |
Chemical Identifiers
6393-56-2 | |
130.9 | |
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf | |
magnesium;pentane;chloride |
C5H11ClMg | |
JFWWQYKSQVMLQU-UHFFFAOYSA-M | |
10953547 | |
CCCC[CH2-].[Mg+2].[Cl-] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger