missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentylmagnesium chloride, 1.3M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

CAS 6393-56-2
Molecular Formula C5H11ClMg
Molecular Weight (g/mol) 130.9
InChI Key JFWWQYKSQVMLQU-UHFFFAOYSA-M
Synonym 1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10953547
IUPAC Name magnesium;pentane;chloride
SMILES CCCC[CH2-].[Mg+2].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436211000
SDS View Documents
Thermo Fisher Scientific
AC436211000
100mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436218000
SDS View Documents
Thermo Fisher Scientific
AC436218000
800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

6393-56-2
130.9
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf
magnesium;pentane;chloride
C5H11ClMg
JFWWQYKSQVMLQU-UHFFFAOYSA-M
10953547
CCCC[CH2-].[Mg+2].[Cl-]

Specifications

0.9670g/mL
AcroSeal™ Glass Bottle
65.0°C
100mL
C5H11ClMg
Solubility in water: reacts.
CCCC[CH2-].[Mg+2].[Cl-]
130.9
130.9
Pentylmagnesium chloride
−17°C
0.967
Undesignated
6393-56-2,109-99-9
1-pentylmagnesium chloride, 1-pentylmagnesium chloride, 1m in methf, chloro pentyl magnesium, n-pentylmagnesium chloride, ohljpymgjfinna-uhfffaoysa-m, pentylmagnesium chloride, pentylmagnesium chloride solution, pentylmagnesium chloride solution, 2.0 m in thf
JFWWQYKSQVMLQU-UHFFFAOYSA-M
magnesium;pentane;chloride
10953547
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

SDS