Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentaphenylcyclopentadiene, 99%

$59.38 - $435.46

Chemical Identifiers

Chemical Name or Material Pentaphenylcyclopentadiene
CAS 2519-10-0
Molecular Formula C35H26
Formula Weight 446.59
MDL Number MFCD09909572
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2722303

  SDS
alfa aesar™
H2722303
1g Each for $144.50

AAH2722306

  SDS
alfa aesar™
H2722306
5g Each for $435.46

AAH27223MD

  SDS
alfa aesar™
H27223MD
250mg Each for $59.38
Description & Specifications

Specifications

Formula Weight 446.59
CAS 2519-10-0
Assay 99%
Quantity 1g
Molecular Formula C35H26
Melting Point 254° to 257°C
MDL Number MFCD09909572
Chemical Name or Material Pentaphenylcyclopentadiene

Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

Solubility
Soluble in pyridine 5 mg/ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.