Please login to your online account to display your discounted pricing

Pentane, ULTRA RESI-ANALYZED™, J.T.Baker™

High purity solvents and salts developed for organic residue extraction/concentration procedures

$209.90 - $1,535.20

Chemical Identifiers

Chemical Name or Material Pentane
CAS 109-66-0
Purity Grade ULTRA RESI-ANALYZED™
Molecular Formula CH3(CH2)3CH3
Formula Weight 72.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003534

 
avantor performance
933302
Narrow Mouth Amber Glass Bottle 1L Each for $209.90

02003538

 
avantor performance
933303
Narrow Mouth Amber Glass Bottle 4L Each for $1,535.20
Description & Specifications

Specifications

Formula Weight 72.15
Flash Point −40.00°C(-40°F)
Chemical Name or Material Pentane
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.63kg
Purity Grade ULTRA RESI-ANALYZED™
Molecular Formula CH3(CH2)3CH3
CAS 109-66-0
Quantity 1L

    For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use