Learn More
Pentane, HPLC Grade, 99+% (n-Pentane, 95% min), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H27427AU
Description
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
Solubility
Miscible with water.
Notes
Stable under normal ambient and anticipated storage and handling conditions of temperature. Avoidable conditions are heat, sparks, fire, and oxidizing agents.
Specifications
Pentane | |
109-66-0 | |
36°C | |
−49°C (−57°F) | |
1.358 | |
MFCD00009498 | |
14,7116 | |
Miscible with water. | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
≥99% |
-130°C | |
0.626 | |
1000 mL | |
Nearly Odorless | |
C5H12 | |
UN1265 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 | |
HPLC |
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Safety and Handling
GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
H225-H304-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
EINECSNumber : 203-692-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only