Please login to your online account to display your discounted pricing

Pentane Anhydrous, DriSolv™, EMD Millipore

Septum-Sealed Bottle

$217.11 - $217.11

Chemical Identifiers

Chemical Name or Material Pentane Anhydrous
CAS 109-66-0
Molecular Formula C5H12
Formula Weight 72.15
UN Number 1265
Synonym n-Pentane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MPX01716

 
emd millipore corporation
PX01716
Glass Bottle 1L Each for $217.11
Description & Specifications

Specifications

Boiling Point 36.06°C
Melting Point −129.6°C
Assay ≥99%
Formula Weight 72.15
Quantity 1L
Storage Information Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Chemical Name or Material Pentane Anhydrous
Ignition Point 260°C (500°F)
Odor Gas
Packaging Glass Bottle
Vapor Pressure 59 kPa (442.83mm Hg) [20°C]
Flash Point Closed cup: <-40°C (<-40°F)
Physical Form Liquid
Color Colorless
Molecular Formula C5H12
CAS 109-66-0
UN Number 1265
Synonym n-Pentane
Solubility Information Insoluble in water
Description Packed under Nitrogen and sealed with a septum.