Please login to your online account to display your discounted pricing

Pentane, 98% min., OmniSolv™, EMD Millipore

$373.26 - $373.26

Chemical Identifiers

Chemical Name or Material Pentane
CAS 109-66-0
Physical Form Liquid
Assay Percent Range 98% min.
Molecular Formula C5H12
Linear Formula CH3(CH2)3CH3
Formula Weight 72.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX01671

 
emd millipore corporation
PX01671
4L Each for $373.26
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 72.15
Color Clear colorless
Quantity 4L
Density 0.63g/cm3 (20°C)
Assay Percent Range 98% min.
Chemical Name or Material Pentane
CAS 109-66-0
Molecular Formula C5H12
Vapor Pressure 570 hPa (20°C)
Ignition Point 285°C
Explosion Limit 1.4 to 8%(V)
Linear Formula CH3(CH2)3CH3
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C