Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentamethylenebis(triphenylphosphonium bromide), 98+%

$54.39 - $176.30

Chemical Identifiers

Chemical Name or Material Pentamethylenebis(triphenylphosphonium bromide)
CAS 22884-31-7
Molecular Formula Br(C6H5)3P(CH2)5P(C6H5)3Br
Formula Weight 754.54
MDL Number MFCD00051887
Beilstein 3875042
Synonym 1,5-Bis(triphenylphosphonio)pentane dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0099706

  SDS
alfa aesar™
L0099706
5g Each for $54.39

AAL0099714

  SDS
alfa aesar™
L0099714
25g Each for $176.30
Description & Specifications

Specifications

Formula Weight 754.54
Assay ≥98%
CAS 22884-31-7
Chemical Name or Material Pentamethylenebis(triphenylphosphonium bromide)
Molecular Formula Br(C6H5)3P(CH2)5P(C6H5)3Br
MDL Number MFCD00051887
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Synonym 1,5-Bis(triphenylphosphonio)pentane dibromide
Melting Point 253° to 260°C
Quantity 5g
Beilstein 3875042

Applications
Pentamethylenebis(triphenylphosphonium bromide), is used as an intermediate in Pharmaceutical research.

Solubility
It is soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. Moisture sensitive. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.