Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentamethylcyclopentadienylzirconium trichloride, 99%

$126.88 - $464.08

Chemical Identifiers

Chemical Name or Material Pentamethylcyclopentadienylzirconium trichloride
CAS 75181-07-6
Formula Weight 332.81
MDL Number MFCD00070444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3955103

  SDS
alfa aesar™
3955103
1g Each for $126.88

AA3955106

  SDS
alfa aesar™
3955106
5g Each for $464.08
Description & Specifications

Specifications

Formula Weight 332.81
Packaging Ampule
Chemical Name or Material Pentamethylcyclopentadienylzirconium trichloride
Color Yellow to yellow-green
Quantity 1g
CAS 75181-07-6
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sensitivity Moisture Sensitive
Melting Point 230°C (dec.)
MDL Number MFCD00070444
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.