missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentamethylcyclopentadienylhafnium trichloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 35349-0050

 View more versions of this product

Catalog No. AC353490050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Pentamethylcyclopentadienylhafnium trichloride
5g
Authentic
Crystals or Crystalline Powder
97%
420.07
Purple
75181-08-7
Glass bottle
≥96.0%
C10 H15 Cl3 Hf
MFCD00070461

Chemical Identifiers

75181-08-7
C10 H15 Cl3 Hf
MFCD00070461
≥96.0%
97%
420.07
Pentamethylcyclopentadienylhafnium trichloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE