Please login to your online account to display your discounted pricing

Pentamethylcyclopentadienylhafnium trichloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Pentamethylcyclopentadienylhafnium trichloride
CAS 75181-08-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10 H15 Cl3 Hf
Formula Weight 420.07
MDL Number MFCD00070461
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353490050

  SDS
thermo fisher scientific
35349-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystals or Crystalline Powder
Formula Weight 420.07
Quantity 5g
Color Purple
Chemical Name or Material Pentamethylcyclopentadienylhafnium trichloride
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10 H15 Cl3 Hf
MDL Number MFCD00070461
CAS 75181-08-7
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic