Please login to your online account to display your discounted pricing

(Pentamethylcyclopentadienyl)iridium(III) Chloride dimer, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377952500

 View more versions of this product

Catalog No. AC377952500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex Ir)
Physical Form Fine Crystalline Powder
Formula Weight 796.73
Quantity 250mg
Color Orange
Merck Index 15, 7226
CAS Max % 100.0
Chemical Name or Material (Pentamethylcyclopentadienyl)iridium(III) chloride dimer
Assay Percent Range 99%
MDL Number MFCD00075435
Health Hazard 1 Warning
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer
CAS Min % 98.5
Molecular Formula C20H30Cl4Ir2
CAS 12354-84-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (Pentamethylcyclopentadienyl)iridium(III) chloride dimer
CAS 12354-84-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (ex Ir)
Assay Percent Range 99%
Molecular Formula C20H30Cl4Ir2
Formula Weight 796.73
MDL Number MFCD00075435
Synonym Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents