missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentamethylbenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1473306

 View more versions of this product

Catalog No. AAA1473306



Specifications

Specifications

Pentamethylbenzonitrile
5g
C12H15N
UN3439
NCDPYDGGUAKABA-UHFFFAOYSA-N
2,3,4,5,6-pentamethylbenzonitrile
78824
98%
158°C to 160°C
5144-10-5
MFCD00015450
pentamethylbenzonitrile, 6-aminobutyric acid, pentamethylbenzonitril, acmc-1axcs, benzonitrile,2,3,4,5,6-pentamethyl, 2,3,4,5,6-pentamethylbenzenecarbonitrile
CC1=C(C(=C(C(=C1C)C)C#N)C)C
173.26
173.26

Chemical Identifiers

5144-10-5
173.26
NCDPYDGGUAKABA-UHFFFAOYSA-N
78824
CC1=C(C(=C(C(=C1C)C)C#N)C)C
C12H15N
MFCD00015450
pentamethylbenzonitrile, 6-aminobutyric acid, pentamethylbenzonitril, acmc-1axcs, benzonitrile,2,3,4,5,6-pentamethyl, 2,3,4,5,6-pentamethylbenzenecarbonitrile
2,3,4,5,6-pentamethylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents