missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentamethylbenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1473303

 View more versions of this product

Catalog No. AAA1473303



Specifications

Specifications

Pentamethylbenzonitrile
1g
C5H8N2O
UN3439
VOTFXESXPPEARL-UHFFFAOYSA-N
3-methyl-4,5-dihydro-1H-pyridazin-6-one
78834
98%
158°C to 160°C
5157-08-4
MFCD00015450
6-methyl-4,5-dihydropyridazin-3 2h-one, 4,5-dihydro-6-methylpyridazin-3 2h-one, 6-methyl-2,3,4,5-tetrahydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dihydro-6-methyl, 4,5-dihydro-6-methyl-3 2h-pyridazinone, 6-methyl-4,5-dihydro-2h-pyridazin-3-one, 6-methyl-4,5-dihydro-3 2h-pyridazinone, wln: t6nmv futj f1, 6-methyl-2,4,5-trihydropyridazin-3-one, 3 2h-pyridazinone,5-dihydro-6-methyl
CC1=NNC(=O)CC1
112.13
173.26

Chemical Identifiers

5157-08-4
112.13
VOTFXESXPPEARL-UHFFFAOYSA-N
78834
CC1=NNC(=O)CC1
C5H8N2O
MFCD00015450
6-methyl-4,5-dihydropyridazin-3 2h-one, 4,5-dihydro-6-methylpyridazin-3 2h-one, 6-methyl-2,3,4,5-tetrahydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dihydro-6-methyl, 4,5-dihydro-6-methyl-3 2h-pyridazinone, 6-methyl-4,5-dihydro-2h-pyridazin-3-one, 6-methyl-4,5-dihydro-3 2h-pyridazinone, wln: t6nmv futj f1, 6-methyl-2,4,5-trihydropyridazin-3-one, 3 2h-pyridazinone,5-dihydro-6-methyl
3-methyl-4,5-dihydro-1H-pyridazin-6-one
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents