Please login to your online account to display your discounted pricing

Pentamethylbenzaldehyde, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  335510050

 View more versions of this product

Catalog No. AC335510050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Formula Weight 176.26
Physical Form Crystalline Powder, Crystals or Flakes
Quantity 5g
Color Light yellow to beige
Melting Point 147° to 153°C
Health Hazard 1 AIR SENSITIVE
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Pentamethylbenzaldehyde
MDL Number MFCD00015560
Assay Percent Range 96%
CAS 17432-38-1
Name Note 96%
Molecular Formula C12H16O
Linear Formula (CH3)5C6C(O)H

Specifications

Chemical Name or Material Pentamethylbenzaldehyde
Name Note 96%
CAS 17432-38-1
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12H16O
Linear Formula (CH3)5C6C(O)H
Formula Weight 176.26
MDL Number MFCD00015560
Accessories

AIR SENSITIVE

SDS & Safety Documents