Please login to your online account to display your discounted pricing

Pentamethylbenzaldehyde, 96%, ACROS Organics™

$101.78 - $101.78

Chemical Identifiers

Chemical Name or Material Pentamethylbenzaldehyde
Name Note 96%
CAS 17432-38-1
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12H16O
Linear Formula (CH3)5C6C(O)H
Formula Weight 176.26
MDL Number MFCD00015560
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335510010

  SDS
thermo fisher scientific
335510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC335510050

  SDS
thermo fisher scientific
335510050
Glass bottle 5g Each for $101.78
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Powder, Crystals or Flakes
Formula Weight 176.26
Quantity 1g
Color Light yellow to beige
Melting Point 147° to 153°C
Chemical Name or Material Pentamethylbenzaldehyde
Packaging Glass bottle
Name Note 96%
Assay Percent Range 96%
Molecular Formula C12H16O
Linear Formula (CH3)5C6C(O)H
MDL Number MFCD00015560
CAS 17432-38-1
Health Hazard 1 AIR SENSITIVE
Infrared Spectrum Authentic
Safety and Handling

AIR SENSITIVE