Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentakis(diethylamino)niobium(V), 99.9% (metals basis), mixture of complexes

Nitride film precursor

Chemical Identifiers

Chemical Name or Material Pentakis(diethylamino)niobium(V)
CAS 25169-05-5
Name Note Mixture of complexes
Linear Formula Nb[N(C2H5)2]5
Formula Weight 453.56
MDL Number MFCD00798536
UN Number UN3267
Synonym Niobium diethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4044527

  SDS
alfa aesar™
4044527
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40445ME

  SDS
alfa aesar™
40445ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 453.56
CAS 25169-05-5
Assay 99.9%
MDL Number MFCD00798536
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
Odor Amine-like
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Physical Form Liquid
Boiling Point 120°C (0.1mmHg)
Name Note Mixture of complexes
Sensitivity Moisture Sensitive
Synonym Niobium diethylamide
UN Number UN3267
Color Dark
Linear Formula Nb[N(C2H5)2]5
Assay Percent Notes (metals basis)
Quantity 2.5g
Chemical Name or Material Pentakis(diethylamino)niobium(V)
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.