Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluorothiophenol, 97%

Chemical Identifiers

Chemical Name or Material Pentafluorothiophenol
CAS 771-62-0
Molecular Formula C6HF5S
Formula Weight 200.13
MDL Number MFCD00004828
UN Number UN2920
Beilstein 1876292
Synonym Pentafluorobenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2232306

  SDS
alfa aesar™
B2232306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2232314

  SDS
alfa aesar™
B2232314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.13
Synonym Pentafluorobenzenethiol
Odor Stench
Chemical Name or Material Pentafluorothiophenol
MDL Number MFCD00004828
Beilstein 1876292
Melting Point −24°C
Assay 97%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 143°C
Health Hazard 1 Danger
CAS 771-62-0
UN Number UN2920
Quantity 5g
Flash Point 51°C (123°F)
RTECS Number DC1940000
Molecular Formula C6HF5S

Applications
Pentafluorothiophenol is used as a fluorinated building block

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.