missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluoropyridine, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 169990050

 View more versions of this product

Catalog No. AC169990050



Description

Description

Specifications

Specifications

Pentafluoropyridine
23°C
Glass bottle
1.54
Colorless
99+%
99+%
(C5N)F5
2,3,4,5,6-pentafluoro-pyridine, acmc-1bl6p, pentafluoropyridine, pentafluorpyridin, perfluoropyridine, pubchem9299, pyridine, pentafluoro
C1(=C(C(=NC(=C1F)F)F)F)F
169.05
169.05
99+%
1.5400g/mL
Authentic
1.3850 to 1.387
84°C
5g
700-16-3
C5F5N
MFCD00006225
XTGOWLIKIQLYRG-UHFFFAOYSA-N
2,3,4,5,6-pentafluoropyridine
69690
Liquid

Chemical Identifiers

700-16-3
169.05
XTGOWLIKIQLYRG-UHFFFAOYSA-N
69690
C1(=C(C(=NC(=C1F)F)F)F)F
C5F5N
MFCD00006225
2,3,4,5,6-pentafluoro-pyridine, acmc-1bl6p, pentafluoropyridine, pentafluorpyridin, perfluoropyridine, pubchem9299, pyridine, pentafluoro
2,3,4,5,6-pentafluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 211-839-9

SDS Documents
Product Certifications