Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluoropropionic anhydride, 98%

Chemical Identifiers

Chemical Name or Material Pentafluoropropionic anhydride
CAS 356-42-3
Molecular Formula (CF3CF2CO)2O
Formula Weight 310.04
MDL Number MFCD00000429
UN Number UN3265
Beilstein 1806446
Synonym Perfluoropropionic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1447214

  SDS
alfa aesar™
A1447214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1447206

  SDS
alfa aesar™
A1447206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1447222

  SDS
alfa aesar™
A1447222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.04
Beilstein 1806446
UN Number UN3265
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula (CF3CF2CO)2O
Synonym Perfluoropropionic anhydride
Boiling Point 73° to 74°C
MDL Number MFCD00000429
Quantity 25g
TSCA TSCA
Health Hazard 1 Danger
Chemical Name or Material Pentafluoropropionic anhydride
Assay 98%
CAS 356-42-3
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Applications
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

Solubility
Reacts with water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.