Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluoropropionic acid, 97%

Chemical Identifiers

Chemical Name or Material Pentafluoropropionic acid
CAS 422-64-0
Molecular Formula CF3CF2CO2H
Formula Weight 164.03
MDL Number MFCD00004170
UN Number UN3265
Beilstein 1773387
Synonym Perfluoropropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1279114

 
alfa aesar™
A1279114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1279122

 
alfa aesar™
A1279122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1279136

 
alfa aesar™
A1279136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.03
Chemical Name or Material Pentafluoropropionic acid
TSCA TSCA
Assay 97%
Molecular Formula CF3CF2CO2H
MDL Number MFCD00004170
CAS 422-64-0
UN Number UN3265
Beilstein 1773387
Synonym Perfluoropropionic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECS Number UF6475000
Odor Rancid
Boiling Point 95° to 97°C
Quantity 25g

Applications
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.