Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluoropropionaldehyde hydrate, tech. 90%

$91.73 - $338.64

Chemical Identifiers

Chemical Name or Material Pentafluoropropionaldehyde hydrate
CAS 422-63-9
Purity Grade Technical Grade
Molecular Formula C3H3F5O2·xH2O
Linear Formula CF3CF2CH(OH)2·xH2O
Formula Weight 166.05 (Anhydrous)
MDL Number MFCD00792902
Beilstein 1705233
Synonym Pentafluoropropanal hydrate; 2,2,3,3,3-Pentafluoro-1,1-propanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0081006

 
alfa aesar™
L0081006
5g Each for $91.73

AAL0081014

 
alfa aesar™
L0081014
25g Each for $338.64
Description & Specifications

Specifications

Formula Weight 166.05 (Anhydrous)
MDL Number MFCD00792902
CAS 422-63-9
Chemical Name or Material Pentafluoropropionaldehyde hydrate
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Melting Point 50° to 54°C
Molecular Formula C3H3F5O2·xH2O
Quantity 5g
Linear Formula CF3CF2CH(OH)2·xH2O
Beilstein 1705233
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Pentafluoropropanal hydrate; 2,2,3,3,3-Pentafluoro-1,1-propanediol
RTECS Number TZ0970000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.