Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluorophenylhydrazine, 97%

$44.85 - $491.70

Chemical Identifiers

Chemical Name or Material Pentafluorophenylhydrazine
CAS 828-73-9
Molecular Formula C6H3F5N2
Formula Weight 198.09g/mol
MDL Number MFCD00007574
Beilstein 747800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1330206

 
alfa aesar™
A13302-06
5g Each for $44.85

AAA1330222

 
alfa aesar™
A13302-22
100g Each for $491.70
Description & Specifications

Specifications

Melting Point 73° to 78°C
Quantity 5g
Chemical Name or Material Pentafluorophenylhydrazine
CAS 828-73-9
Assay 97%
Molecular Formula C6H3F5N2
Formula Weight 198.09g/mol
MDL Number MFCD00007574
Beilstein 747800
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MV8325000

Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.