missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America P21881G

 View more versions of this product

Catalog No. P21881G



Specifications

Specifications

Pentafluorophenyl Trifluoromethanesulfonate
Yellow
60129-85-3
C7F8O3S
Trifluoromethanesulfonic Acid Pentafluorophenyl Ester, Pentafluorophenyl Triflate
C1(=C(C(=C(C(=C1F)F)F)F)F)OS(=O)(=O)C(F)(F)F
316.121
316.12
40°C
1g
≥97.0% (GC)
MFCD07784319
OTTBRWDUQHDNBY-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) trifluoromethanesulfonate
10041425
Clear Liquid at 20°C

Chemical Identifiers

60129-85-3
316.121
OTTBRWDUQHDNBY-UHFFFAOYSA-N
10041425
C7F8O3S
MFCD07784319
Trifluoromethanesulfonic Acid Pentafluorophenyl Ester, Pentafluorophenyl Triflate
Safety and Handling

Safety and Handling

TSCA : No

Documents