missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluorophenyl trifluoroacetate, 98+%

Manufacturer:  Alfa Aesar™ H5452914

 View more versions of this product

Catalog No. AAH5452914



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 122.5°Cat760mmHg.

Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Pentafluorophenyl trifluoroacetate
122°C to 123°C
14533-84-7
MFCD00134438
2003848
VCQURUZYYSOUHP-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
4327891
≥98%
52°C (126°F)
25g
C8F8O2
UN3272
acetic acid, trifluoro-, pentafluorophenyl ester
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)C(F)(F)F
280.073
280.07

Chemical Identifiers

14533-84-7
280.073
VCQURUZYYSOUHP-UHFFFAOYSA-N
4327891
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)C(F)(F)F
C8F8O2
MFCD00134438
acetic acid, trifluoro-, pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Documents