missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluorophenyl trifluoroacetate, 98+%

Manufacturer:  Alfa Aesar™ H5452906

 View more versions of this product

Catalog No. AAH5452906



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 122.5°Cat760mmHg.

Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Pentafluorophenyl trifluoroacetate
52°C (126°F)
Moisture sensitive
5g
C8F8O2
UN3272
acetic acid, trifluoro-, pentafluorophenyl ester
VCQURUZYYSOUHP-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
4327891
≥98%
1.63
1.368
122°C to 123°C
14533-84-7
MFCD00134438
2003848
Soluble in water at 122.5°Cat760mmHg.
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)C(F)(F)F
280.073
280.07

Chemical Identifiers

14533-84-7
280.073
VCQURUZYYSOUHP-UHFFFAOYSA-N
4327891
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)C(F)(F)F
C8F8O2
MFCD00134438
acetic acid, trifluoro-, pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents