missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl pyridine-2-carboxylate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 188837-53-8
Molecular Formula C12H4F5NO2
Molecular Weight (g/mol) 289.16
MDL Number MFCD02916457
InChI Key KFJSLIBBYMXLTG-UHFFFAOYSA-N
Synonym pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 2754830
IUPAC Name (2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC04026CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC04026CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC04026DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC04026DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

188837-53-8
289.16
KFJSLIBBYMXLTG-UHFFFAOYSA-N
2754830
FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F
C12H4F5NO2
MFCD02916457
pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate

Specifications

188837-53-8
100.0
Amber Glass Bottle
MFCD02916457
pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F
289.16
289.16
Pentafluorophenyl pyridine-2-carboxylate
94.5
95%
C12H4F5NO2
250mg
KFJSLIBBYMXLTG-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate
2754830
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'