Learn More
Pentafluorophenyl nicotinate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 848347-44-4 |
---|---|
Molecular Formula | C12H4F5NO2 |
Molecular Weight (g/mol) | 289.161 |
MDL Number | MFCD09064938 |
InChI Key | AXHLJDBUUFUDCF-UHFFFAOYSA-N |
Synonym | pentafluorophenyl nicotinate, pentafluorophenyl pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-3-carboxylate, 3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester |
PubChem CID | 23237920 |
IUPAC Name | (2,3,4,5,6-pentafluorophenyl) pyridine-3-carboxylate |
SMILES | C1=CC(=CN=C1)C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC04126CB
|
Thermo Scientific Chemicals
CC04126CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC04126DA
|
Thermo Scientific Chemicals
CC04126DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
848347-44-4 | |
289.161 | |
AXHLJDBUUFUDCF-UHFFFAOYSA-N | |
23237920 | |
C1=CC(=CN=C1)C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F |
C12H4F5NO2 | |
MFCD09064938 | |
pentafluorophenyl nicotinate, pentafluorophenyl pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-3-carboxylate, 3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester | |
(2,3,4,5,6-pentafluorophenyl) pyridine-3-carboxylate |
Specifications
848347-44-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD09064938 | |
pentafluorophenyl nicotinate, pentafluorophenyl pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-3-carboxylate, 3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester | |
C1=CC(=CN=C1)C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F | |
289.161 | |
289.16 | |
Pentafluorophenyl nicotinate |
96.5 | |
97% | |
C12H4F5NO2 | |
250mg | |
AXHLJDBUUFUDCF-UHFFFAOYSA-N | |
(2,3,4,5,6-pentafluorophenyl) pyridine-3-carboxylate | |
23237920 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning