missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl Methacrylate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America P22895G

 View more versions of this product

Catalog No. P22895G



Specifications

Specifications

Pentafluorophenyl Methacrylate (stabilized with MEHQ)
5g
C10H5F5O2
Methacrylic Acid Pentafluorophenyl Ester
CC(=C)C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F
252.14
252.14
≥97.0% (GC)
Yellow
13642-97-2
MFCD00042332
NIJWSVFNELSKMF-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 2-methylprop-2-enoate
193480
Clear Liquid at 20°C

Chemical Identifiers

13642-97-2
252.14
NIJWSVFNELSKMF-UHFFFAOYSA-N
193480
CC(=C)C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F
C10H5F5O2
MFCD00042332
Methacrylic Acid Pentafluorophenyl Ester
(2,3,4,5,6-pentafluorophenyl) 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents