missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl isothiocyanate, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1187406

 View more versions of this product

Catalog No. AAL1187406



Description

Description

Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Specifications

Pentafluorophenyl isothiocyanate
1.56
C7F5NS
MFCD00041042
UN2922
Moisture sensitive
NGNKMRBGZPDABE-UHFFFAOYSA-N
1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
123452
96%
35923-79-6
109°C to 110°C (35 mmHg)
1.5378
5 g
2736207
pentafluorophenyl isothiocyanate, perfluorophenyl isothiocyanate, pentafluorophenylisothiocyanate, benzene, pentafluoroisothiocyanato, 2,3,4,5,6-pentafluorophenylisothiocyanate, 2,3,4,5,6-pentafluorophenyl isothiocyanate, acmc-20ajyt, pubchem5009, pentafluorophenyl isothiocyanate 1g, 2,3,4,5,6-pentafluorobenzenisothiocyanate
C(=NC1=C(C(=C(C(=C1F)F)F)F)F)=S
225.136
225.14

Chemical Identifiers

35923-79-6
225.136
NGNKMRBGZPDABE-UHFFFAOYSA-N
123452
C(=NC1=C(C(=C(C(=C1F)F)F)F)F)=S
C7F5NS
MFCD00041042
pentafluorophenyl isothiocyanate, perfluorophenyl isothiocyanate, pentafluorophenylisothiocyanate, benzene, pentafluoroisothiocyanato, 2,3,4,5,6-pentafluorophenylisothiocyanate, 2,3,4,5,6-pentafluorophenyl isothiocyanate, acmc-20ajyt, pubchem5009, pentafluorophenyl isothiocyanate 1g, 2,3,4,5,6-pentafluorobenzenisothiocyanate
1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 252-791-9

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only