missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentafluorophenyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
$172.54 - $640.38
Chemical Identifiers
CAS | 71195-85-2 |
---|---|
Molecular Formula | C9H3F5O2 |
Molecular Weight (g/mol) | 238.11 |
MDL Number | MFCD00042330 |
InChI Key | RFOWDPMCXHVGET-UHFFFAOYSA-N |
Synonym | Acrylic Acid Pentafluorophenyl Ester |
PubChem CID | 2775973 |
IUPAC Name | 2,3,4,5,6-pentafluorophenyl prop-2-enoate |
SMILES | FC1=C(F)C(F)=C(OC(=O)C=C)C(F)=C1F |
Chemical Identifiers
71195-85-2 | |
238.11 | |
RFOWDPMCXHVGET-UHFFFAOYSA-N | |
2775973 | |
FC1=C(F)C(F)=C(OC(=O)C=C)C(F)=C1F |
C9H3F5O2 | |
MFCD00042330 | |
Acrylic Acid Pentafluorophenyl Ester | |
2,3,4,5,6-pentafluorophenyl prop-2-enoate |
Specifications
71195-85-2 | |
197°C | |
MFCD00042330 | |
Acrylic Acid Pentafluorophenyl Ester | |
FC1=C(F)C(F)=C(OC(=O)C=C)C(F)=C1F | |
238.11 | |
238.11 | |
Liquid |
Colorless | |
C9H3F5O2 | |
1 g | |
RFOWDPMCXHVGET-UHFFFAOYSA-N | |
2,3,4,5,6-pentafluorophenyl prop-2-enoate | |
2775973 | |
≥98.0% (GC) | |
Pentafluorophenyl Acrylate (stabilized with MEHQ) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator