missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC60626DA

 View more versions of this product

Catalog No. CC60626DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

368.26
97%
Amber Glass Bottle
97%
C17H9F5N2O2
MFCD09065010
1g
100.0
910037-11-5
Pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate
96.5

Chemical Identifiers

910037-11-5
100.0
C17H9F5N2O2
MFCD09065010
97%
96.5
97%
368.26
Pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning