missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, 97%, Maybridge

Chemical Identifiers

CAS 906352-58-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H5F5N2O2S
Formula Weight 372.27
MDL Number MFCD09025821
Chemical Name or Material Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC13726DA SDS Fisher Scientific Company
CC13726DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC13726CB SDS Fisher Scientific Company
CC13726CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

906352-58-7
100.0
C15H5F5N2O2S
MFCD09025821
97%
96.5
97%
372.27
Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate

Specifications

1g
96.5
97%
372.27
Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate
906352-58-7
100.0
C15H5F5N2O2S
MFCD09025821
97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.