Please login to your online account to display your discounted pricing

Pentafluorophenyl 2-morpholino-5-(trifluoromethyl)benzoate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC57826DA

 View more versions of this product

Catalog No. CC57826DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 441.27
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Pentafluorophenyl 2-morpholino-5-(trifluoromethyl)benzoate
Assay Percent Range 97%
Molecular Formula C18H11F8NO3
MDL Number MFCD09025890
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
CAS 898289-52-6
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Pentafluorophenyl 2-morpholino-5-(trifluoromethyl)benzoate
CAS 898289-52-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H11F8NO3
Formula Weight 441.27
MDL Number MFCD09025890
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents