missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate, 97%, Maybridge

Chemical Identifiers

CAS 898288-88-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H5F5N2O2
Formula Weight 292.16
MDL Number MFCD09025820
Chemical Name or Material Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC03626CB SDS Fisher Scientific Company
CC03626CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC03626DA SDS Fisher Scientific Company
CC03626DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

898288-88-5
100.0
C11H5F5N2O2
MFCD09025820
97%
96.5
97%
292.16
Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate

Specifications

250mg
96.5
Amber Glass Bottle
C11H5F5N2O2
MFCD09025820
97%
898288-88-5
100.0
97%
292.16
Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning