Learn More
Pentafluorobenzoyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 147121000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Acylation reagentSpecifications
Pentafluorobenzoyl chloride | |
1.6000g/mL | |
97.5% min. (GC) | |
C7ClF5O | |
C6F5COCl | |
100g | |
pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate | |
MYHOHFDYWMPGJY-UHFFFAOYSA-N | |
pentafluorobenzoyl chloride | |
75256 | |
230.52 |
2251-50-5 | |
Authentic | |
Glass Bottle | |
1.4520 to 1.4540 | |
MFCD00000657 | |
1.6 | |
Solubility in water: decomposes. | |
FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F | |
230.52 | |
CHEBI:39425 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 218-842-4
RUO â Research Use Only