missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentafluorobenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1242006

 View more versions of this product

Catalog No. AAA1242006


 

Description

Description

Applications
A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Water, Alcohols, Strong bases.
Specifications

Specifications

Pentafluorobenzoyl chloride
98%
C7ClF5O
MFCD00000657
2054397
MYHOHFDYWMPGJY-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzoyl chloride
75256
230.52
2251-50-5
158°C to 159°C
5g
UN3265
Perfluorobenzoyl chloride
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)Cl
230.518
CHEBI:39425

Chemical Identifiers

2251-50-5
230.518
MYHOHFDYWMPGJY-UHFFFAOYSA-N
75256
C7ClF5O
MFCD00000657
Perfluorobenzoyl chloride
CHEBI:39425
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents