Learn More
Pentafluorobenzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1239614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pentafluorobenzoic acid | |
100°C to 104°C | |
C7HF5O2 | |
25 g | |
pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017 | |
OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F | |
212.08 | |
CHEBI:46796 | |
99% |
602-94-8 | |
219°C to 221°C | |
MFCD00002406 | |
2054395 | |
YZERDTREOUSUHF-UHFFFAOYSA-N | |
2,3,4,5,6-pentafluorobenzoic acid | |
11770 | |
212.07 |
Chemical Identifiers
602-94-8 | |
212.08 | |
YZERDTREOUSUHF-UHFFFAOYSA-N | |
11770 | |
2,3,4,5,6-pentafluorobenzoic acid |
C7HF5O2 | |
MFCD00002406 | |
pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017 | |
CHEBI:46796 | |
OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-026-6
RTECSNumber : DH6195000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only