missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorobenzenesulfonyl chloride, 99%, ACROS Organics™

Manufacturer:  Acros Organics 147110010

 View more versions of this product

Catalog No. AC147110010



Specifications

Specifications

Pentafluorobenzenesulfonyl chloride
Glass bottle
99%
99%
C6F5SO2Cl
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
266.566
266.57
1.7960g/mL
1g
832-53-1
C6ClF5O2S
MFCD00007427
UOJCTEGNHXRPKO-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
70026

Chemical Identifiers

832-53-1
266.566
UOJCTEGNHXRPKO-UHFFFAOYSA-N
70026
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
C6ClF5O2S
MFCD00007427
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 212-620-0

SDS Documents