missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluoro(phenoxy)cyclotriphosphazene, TCI America™

Manufacturer:  TCI America P22865G

 View more versions of this product

Catalog No. P22865G



Specifications

Specifications

Pentafluoro(phenoxy)cyclotriphosphazene
Colorless
33027-68-8
C6H5F5N3OP3
2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(F)F)(F)F)F
323.039
323.04
46°C
5g
≥98.0% (GC)
1760
XNZZEQCBAGUFMT-UHFFFAOYSA-N
2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
23419251
Clear Liquid at 20°C

Chemical Identifiers

33027-68-8
323.039
2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine
2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
C6H5F5N3OP3
XNZZEQCBAGUFMT-UHFFFAOYSA-N
23419251
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(F)F)(F)F)F
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6917

TSCA : No

Recommended Storage : Refrigerator

Documents