Please login to your online account to display your discounted pricing

Pentaethylenehexamine, Tech., 30.5% nitrogen, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC320290010

 View more versions of this product

Catalog No. AC320290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.44
Purity Grade Technical Grade
Assay ≥30.5% (NITROGEN)
Physical Form Liquid
Quantity 1L
Color Clear yellow
RTECS Number RZ2680000
TSCA TSCA
EINECS Number 223-775-9
Density 0.95g/mL
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
MDL Number MFCD00008167
Water 0.5% max. (K.F.)
Refractive Index 1.5086 to 1.5106 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula HN(CHCHNH)4CHCHNH
CAS 4067-16-7
Chemical Name or Material Pentaethylenehexamine
Molecular Formula C10H28N6

Specifications

Chemical Name or Material Pentaethylenehexamine
CAS 4067-16-7
Purity Grade Technical Grade
Assay ≥30.5% (NITROGEN)
Molecular Formula C10H28N6
Linear Formula HN(CHCHNH)4CHCHNH
Formula Weight 232.44
MDL Number MFCD00008167
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents