Please login to your online account to display your discounted pricing

Pentaethylenehexamine, Tech., 30.5% nitrogen, ACROS Organics™

$53.14 - $53.14

Chemical Identifiers

Chemical Name or Material Pentaethylenehexamine
CAS 4067-16-7
Purity Grade Technical Grade
Assay ≥30.5% (NITROGEN)
Molecular Formula C10H28N6
Linear Formula HN(CHCHNH)4CHCHNH
Formula Weight 232.44
MDL Number MFCD00008167
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320290010

  SDS
thermo fisher scientific
AC320290010
Glass bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC320292500

  SDS
thermo fisher scientific
320292500
Glass bottle 250mL Each for $53.14
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 232.44
Physical Form Liquid
Assay ≥30.5% (NITROGEN)
Quantity 1L
Color Clear yellow
EINECS Number 223-775-9
TSCA TSCA
RTECS Number RZ2680000
Density 0.95g/mL
Packaging Glass bottle
Water 0.5% max. (K.F.)
MDL Number MFCD00008167
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Chemical Name or Material Pentaethylenehexamine
Refractive Index 1.5086 to 1.5106 (20°C, 589nm)
Molecular Formula C10H28N6
Linear Formula HN(CHCHNH)4CHCHNH
CAS 4067-16-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.