Please login to your online account to display your discounted pricing

Pentaethylenehexamine, Tech., 30.5% nitrogen, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  320292500

 View more versions of this product

Catalog No. AC320292500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥30.5% (NITROGEN)
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 232.44
Quantity 250mL
Color Clear yellow
EINECS Number 223-775-9
TSCA TSCA
RTECS Number RZ2680000
Density 0.95g/mL
Chemical Name or Material Pentaethylenehexamine
Packaging Glass bottle
Refractive Index 1.5086 to 1.5106 (20°C, 589nm)
Water 0.5% max. (K.F.)
Molecular Formula C10H28N6
Linear Formula HN(CHCHNH)4CHCHNH
MDL Number MFCD00008167
CAS 4067-16-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Pentaethylenehexamine
CAS 4067-16-7
Purity Grade Technical Grade
Assay ≥30.5% (NITROGEN)
Molecular Formula C10H28N6
Linear Formula HN(CHCHNH)4CHCHNH
Formula Weight 232.44
MDL Number MFCD00008167
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents