missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentaethylene glycol di-p-toluenesulfonate, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1436903

 View more versions of this product

Catalog No. AAL1436903



Description

Description

Pentaethylene glycol di-p-toluenesulfonate is used in the preparation of calix[4]arenes, calix[5]arenes, and crown ethers. It is also used in the preparation of 3?-formylbenzo-18-crown-6 by undergoing coupling reaction with 2,3-dihydroxybenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaethylene glycol di-p-toluenesulfonate is used in the preparation of calix[4]arenes, calix[5]arenes, and crown ethers. It is also used in the preparation of 3′-formylbenzo-18-crown-6 by undergoing coupling reaction with 2,3-dihydroxybenzaldehyde.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
Specifications

Specifications

Pentaethylene glycol di-p-toluenesulfonate
1.26
C24H34O10S2
MFCD00012204
3599245
3,6,9,12-tetraoxatetradecane-1,14-diyl bis 4-methylbenzenesulfonate, pentaethylene glycol ditosylate, pentaethylene glycol di-p-toluenesulfonate, pentaethylene glycol di p-toluenesulfonate, tos-peg6-tos, 3,6,9,12-tetraoxatetradecane-1,14-diol, bis 4-methylbenzenesulfonate, pentaethylene glycol d p-toluenesulfonate, penta ethylene glycol di-p-toluenesulfonate, 3,6,9,12-tetraoxatetradecane-1,14-diyl ditosylate
BUHGDYPBQWWWQS-UHFFFAOYSA-N
2-[2-[2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
361968
95%
41024-91-3
>110°C (230°F)
1.5225
1 g
Hygroscopic
Not miscible or difficult to mix in water.
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C
546.646
546.66

Chemical Identifiers

41024-91-3
546.646
BUHGDYPBQWWWQS-UHFFFAOYSA-N
361968
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C
C24H34O10S2
MFCD00012204
3,6,9,12-tetraoxatetradecane-1,14-diyl bis 4-methylbenzenesulfonate, pentaethylene glycol ditosylate, pentaethylene glycol di-p-toluenesulfonate, pentaethylene glycol di p-toluenesulfonate, tos-peg6-tos, 3,6,9,12-tetraoxatetradecane-1,14-diol, bis 4-methylbenzenesulfonate, pentaethylene glycol d p-toluenesulfonate, penta ethylene glycol di-p-toluenesulfonate, 3,6,9,12-tetraoxatetradecane-1,14-diyl ditosylate
2-[2-[2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only