missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentaethylene glycol, >98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1013330

 View more versions of this product

Catalog No. AAA1013330



Description

Description

Applications
Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications

Specifications

Pentaethylene glycol
250g
>98%
MFCD00002878
JLFNLZLINWHATN-UHFFFAOYSA-N
2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
62551
238.28
1.126g/mL
4792-15-8
C10H22O6
pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579
C(COCCOCCOCCOCCO)O
238.28
CHEBI:39631

Chemical Identifiers

4792-15-8
238.28
JLFNLZLINWHATN-UHFFFAOYSA-N
62551
2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
C10H22O6
MFCD00002878
pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579
CHEBI:39631
C(COCCOCCOCCOCCO)O
Safety and Handling

Safety and Handling

EINECSNumber : 225-341-4

SDS Documents