Please login to your online account to display your discounted pricing

Pentaethylene glycol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  350730050

 View more versions of this product

Catalog No. AC350730050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 238.28
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
RTECS Number RZ2670000
TSCA TSCA
EINECS Number 225-341-4
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Name Note 98%
Molecular Formula C10H22O6
Assay Percent Range 98%
MDL Number MFCD00002878
Chemical Name or Material Pentaethylene glycol
CAS 4792-15-8

Specifications

Chemical Name or Material Pentaethylene glycol
Name Note 98%
CAS 4792-15-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H22O6
Formula Weight 238.28
MDL Number MFCD00002878
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents