missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentaerythritol triacrylate, stab. with 300-400ppm 4-methoxyphenol

$67.27 - $67.27

Chemical Identifiers

CAS 3524-68-3
Formula Weight 298.28
MDL Number MFCD00009607
Beilstein 6598752
Synonym PETA
Chemical Name or Material Pentaerythritol triacrylate, Stabilized with 300 to 400ppm 4-methoxyphenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1570536 SDS Alfa Aesar™
1570536
500g Each for $67.27
Specifications Description & Specifications

Chemical Identifiers

3524-68-3
MFCD00009607
PETA
298.28
6598752
Pentaerythritol triacrylate, Stabilized with 300 to 400ppm 4-methoxyphenol

Specifications

1.16g/mL
1.484
Liquid
>110° (230°F)
MFCD00009607
6598752
Pentaerythritol triacrylate, Stabilized with 300 to 400ppm 4-methoxyphenol
3524-68-3
Colorless
500g
298.28
Hygroscopic
PETA

Safety and Handling

GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P362-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UD3370000

TSCA : TSCA